1-(3,6-Difluoro-2-hydroxyphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

75590-500MG 75590-1G 75590-5G
77334-142EA 326.51 USD
77334-142 77358-324 77382-232
1-(3,6-Difluoro-2-hydroxyphenyl)-2,2-difluoroethanone ≥95%
1-(3,6-Difluoro-2-hydroxyphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₄F₄O₂
MW: 208.1097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766073
CAS Number: 2901105-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR