1-(6-Bromo-2-fluoro-3-hydroxyphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

75559-500MG 75559-1G 75559-5G
77339-834EA 655.08 USD
77339-834 77358-322 77382-230
1-(6-Bromo-2-fluoro-3-hydroxyphenyl)-2,2-difluoroethanone ≥97%
1-(6-Bromo-2-fluoro-3-hydroxyphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O₂
MW: 269.0153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766064
CAS Number: 2901082-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR