1-(2-Chloro-6-fluoro-3-hydroxyphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

75554-500MG 75554-1G 75554-5G
77335-728EA 363.59 USD
77335-728 77358-320 77382-228
1-(2-Chloro-6-fluoro-3-hydroxyphenyl)-2,2-difluoroethanone ≥95%
1-(2-Chloro-6-fluoro-3-hydroxyphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MW: 224.5643 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766062
CAS Number: 2901065-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR