1-(2-Ethoxy-4-fluorophenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

75547-500MG 75547-1G 75547-5G 75547-25G
77334-140EA 202.91 USD
77334-140 77358-318 77370-152 77390-968
1-(2-Ethoxy-4-fluorophenyl)-2,2-difluoroethanone ≥97%
1-(2-Ethoxy-4-fluorophenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂
MW: 218.1724 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766059
CAS Number: 2901105-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR