4-Butoxy-3-fluoro-2-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

75534-500MG 75534-1G 75534-5G
77335-962EA 370.8 USD
77335-962 77358-316 77382-226
4-Butoxy-3-fluoro-2-formylbenzoic acid ≥97%
4-Butoxy-3-fluoro-2-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃FO₄
MW: 240.2276 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766053
CAS Number: 2807467-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR