1-Bromo-2-iodo-4-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

75512-500MG 75512-1G 75512-5G 75512-25G
77334-136EA 157.59 USD
77334-136 77345-678 77370-148 77390-964
1-Bromo-2-iodo-4-isopropoxybenzene ≥97%
1-Bromo-2-iodo-4-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrIO
MW: 340.9835 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29094052
CAS Number: 1369784-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR