1-Bromo-4-ethoxy-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

75505-500MG 75505-1G 75505-5G 75505-25G
77334-134EA 157.59 USD
77334-134 77345-726 77370-146 77390-962
1-Bromo-4-ethoxy-2-iodobenzene ≥97%
1-Bromo-4-ethoxy-2-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrIO
MW: 326.9569 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766036
CAS Number: 2807471-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR