5-Bromo-1-chloro-2-fluoro-3-isobutoxybenzene ≥97%

Supplier: AOBChem USA

Warning

75496-500MG 75496-1G 75496-5G 75496-25G
77334-132EA 120.51 USD
77334-132 77343-836 77370-144 77382-222
5-Bromo-1-chloro-2-fluoro-3-isobutoxybenzene ≥97%
5-Bromo-1-chloro-2-fluoro-3-isobutoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrClFO
MW: 281.5491 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766027
CAS Number: 2901107-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR