3-Chloro-4-fluoro-5-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

75491-500MG 75491-1G 75491-5G
77336-756EA 393.46 USD
77336-756 77358-310 77382-220
3-Chloro-4-fluoro-5-isopropoxybenzoic acid ≥97%
3-Chloro-4-fluoro-5-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD34766022
CAS Number: 2901085-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR