5-Bromo-1-chloro-2-fluoro-3-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

75489-500MG 75489-1G 75489-5G 75489-25G
77334-130EA 181.28 USD
77334-130 77346-874 77370-142 77390-960
5-Bromo-1-chloro-2-fluoro-3-isopropoxybenzene ≥97%
5-Bromo-1-chloro-2-fluoro-3-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉BrClFO
Storage Temperature: Ambient
MDL Number: MFCD29043894
CAS Number: 1881293-48-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR