4-Bromo-1-isopropyl-1H-pyrazole-5-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

75469-500MG 75469-1G 75469-25G 75469-5G
77334-128EA 305.91 USD
77334-128 77358-308 77390-958 77382-218
4-Bromo-1-isopropyl-1H-pyrazole-5-carbaldehyde ≥95%
4-Bromo-1-isopropyl-1H-pyrazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₉BrN₂O
MW: 217.0631 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26679350
CAS Number: 1517839-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR