4-Bromo-1-isobutyl-1H-pyrazole-5-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

75467-500MG 75467-1G 75467-5G 75467-25G
77334-126EA 204.97 USD
77334-126 77358-306 77370-140 77390-956
4-Bromo-1-isobutyl-1H-pyrazole-5-carbaldehyde ≥97%
4-Bromo-1-isobutyl-1H-pyrazole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₁₁BrN₂O
MW: 231.0897 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31417579
CAS Number: 1784836-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR