5-(Methoxymethyl)-1-methyl-1H-pyrazole-4-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

75463-500MG 75463-1G 75463-5G
77337-570EA 423.33 USD
77337-570 77358-304 77382-216
5-(Methoxymethyl)-1-methyl-1H-pyrazole-4-carboxylic acid ≥95%
5-(Methoxymethyl)-1-methyl-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₃
MW: 170.1659 g/mol
Storage Temperature: Ambient
CAS Number: 1602394-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR