1-Fluoro-4-iodo-2-naphthaldehyde ≥97%

Supplier: AOBChem USA

Warning

75446-500MG 75446-1G 75446-5G
77337-958EA 454.23 USD
77337-958 77358-298 77382-214
1-Fluoro-4-iodo-2-naphthaldehyde ≥97%
1-Fluoro-4-iodo-2-naphthaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₆FIO
MW: 300.0676 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766003
CAS Number: 2807464-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR