(2-Fluoro-6-methoxy-4-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

75435-500MG 75435-1G 75435-5G 75435-25G
77334-122EA 157.59 USD
77334-122 77345-724 77370-136 77390-952
(2-Fluoro-6-methoxy-4-methylphenyl)(methyl)sulfane ≥97%
(2-Fluoro-6-methoxy-4-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁FOS
MW: 186.2464 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765997
CAS Number: 2706784-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR