2,6-Dibromo-3,4-difluorophenol ≥97%

Supplier: AOBChem USA

Warning

75415-1G 75415-5G 75415-25G
77340-916EA 62.83 USD
77340-916 77370-130 77382-208
2,6-Dibromo-3,4-difluorophenol ≥97%
2,6-Dibromo-3,4-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₂Br₂F₂O
MW: 287.8842 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28784357
CAS Number: 1780839-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR