4-Acetyl-2,6-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

75341-500MG 75341-1G 75341-25G 75341-5G
77334-114EA 305.91 USD
77334-114 77358-286 77390-944 77382-204
4-Acetyl-2,6-dimethoxybenzaldehyde ≥97%
4-Acetyl-2,6-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₄
MW: 208.2105 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765962
CAS Number: 2386045-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR