2-(Benzyloxy)-1-bromo-5-fluoro-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

75168-1G 75168-5G 75168-25G
77341-180EA 73.13 USD
77341-180 77370-126 77382-202
2-(Benzyloxy)-1-bromo-5-fluoro-3-methoxybenzene ≥97%
2-(Benzyloxy)-1-bromo-5-fluoro-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO₂
MW: 311.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765838
CAS Number: 2807447-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR