1-Bromo-5-fluoro-2-isobutoxy-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

75166-1G 75166-5G 75166-25G
77341-200EA 75.19 USD
77341-200 77370-124 77382-200
1-Bromo-5-fluoro-2-isobutoxy-3-methoxybenzene ≥97%
1-Bromo-5-fluoro-2-isobutoxy-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrFO₂
MW: 277.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765836
CAS Number: 2807448-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR