4-Fluoro-2-(methylsulfonyl)toluene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoro-1-methyl-2-(methylsulphonyl)benzene

75163-500MG 75163-1G 75163-5G 75163-25G
77334-110EA 112.27 USD
77334-110 77343-316 77370-120 77382-198
4-Fluoro-2-(methylsulfonyl)toluene ≥97%
4-Fluoro-2-(methylsulfonyl)toluene

Precaución: for Research Use Only

Formula: C₈H₉FO₂S
MW: 188.219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09259030
CAS Number: 828270-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR