1-Bromo-3-chloro-2-fluoro-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

75154-500MG 75154-1G 75154-5G 75154-25G
77334-108EA 263.68 USD
77334-108 77358-282 77370-118 77390-940
1-Bromo-3-chloro-2-fluoro-5-methylbenzene ≥97%
1-Bromo-3-chloro-2-fluoro-5-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅BrClF
Storage Temperature: Ambient
MDL Number: MFCD28734048
CAS Number: 1780248-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR