(3-Chloro-2-fluoro-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

75152-1G 75152-5G 75152-25G
77340-914EA 62.83 USD
77340-914 77370-116 77382-196
(3-Chloro-2-fluoro-5-methylphenyl)(methyl)sulfane ≥97%
(3-Chloro-2-fluoro-5-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈ClFS
MW: 190.666 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28752233
CAS Number: 1807272-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR