Methyl 3-chloro-2-fluoro-5-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

75147-500MG 75147-1G 75147-5G
77337-902EA 450.11 USD
77337-902 77358-280 77382-194
Methyl 3-chloro-2-fluoro-5-methylbenzoate ≥97%
Methyl 3-chloro-2-fluoro-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
MW: 202.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28650873
CAS Number: 1782551-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR