3-Chloro-2-fluoro-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

75146-500MG 75146-1G 75146-25G 75146-5G
77334-106EA 300.76 USD
77334-106 77358-278 77390-938 77382-192
3-Chloro-2-fluoro-5-methylbenzoic acid ≥97%
3-Chloro-2-fluoro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD23712072
CAS Number: 1427427-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR