5-Chloro-3-fluoro-2-(2-methoxyethoxy)isonicotinic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
75129-500MG
75129-1G
75129-5G
75129-25G
77334-104EA
120.51
USD
77334-104
77343-834
77370-114
77382-188
5-Chloro-3-fluoro-2-(2-methoxyethoxy)isonicotinic acid ≥97%
5-Chloro-3-fluoro-2-(2-methoxyethoxy)isonicotinic acid
Precaución: for Research Use Only
Formula:
C₉H₉ClFNO₄ MW: 249.6234 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34765819 CAS Number: 2807464-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...