1-(3,5-Difluoro-4-methylphenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

75102-500MG 75102-1G 75102-5G 75102-25G
77334-100EA 136.99 USD
77334-100 77344-638 77370-110 77390-936
1-(3,5-Difluoro-4-methylphenyl)-2,2,2-trifluoroethanone ≥95%
1-(3,5-Difluoro-4-methylphenyl)-2,2,2-trifluoroethanone

Precaución: for Research Use Only

Formula: C₉H₅F₅O
MW: 224.1274 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765798
CAS Number: 2807464-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR