4-Bromo-3-(dibromomethyl)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

75100-500MG 75100-1G 75100-5G 75100-25G
77334-098EA 90.64 USD
77334-098 77342-406 77370-108 77382-184
4-Bromo-3-(dibromomethyl)benzonitrile ≥95%
4-Bromo-3-(dibromomethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₄Br₃N
Storage Temperature: Ambient
MDL Number: MFCD24335261
CAS Number: 160313-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR