3-(Benzyloxy)-5-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

75070-500MG 75070-1G 75070-5G 75070-25G
77334-094EA 208.06 USD
77334-094 77358-274 77370-104 77390-934
3-(Benzyloxy)-5-(trifluoromethoxy)benzaldehyde ≥97%
3-(Benzyloxy)-5-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₁F₃O₃
MW: 296.241 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765783
CAS Number: 1645953-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR