3-(Benzyloxy)-5-(trifluoromethoxy)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

75068-500MG 75068-1G 75068-5G 75068-25G
77334-092EA 182.31 USD
77334-092 77347-038 77370-102 77390-932
3-(Benzyloxy)-5-(trifluoromethoxy)benzoic acid ≥95%
3-(Benzyloxy)-5-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₁F₃O₄
MW: 312.241 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765781
CAS Number: 2807467-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR