4-Bromo-2,5-bis(methylthio)thiophene-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

75062-500MG 75062-1G 75062-5G
77337-342EA 410.97 USD
77337-342 77358-270 77382-178
4-Bromo-2,5-bis(methylthio)thiophene-3-carboxylic acid ≥97%
4-Bromo-2,5-bis(methylthio)thiophene-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₇BrO₂S₃
Storage Temperature: Ambient
CAS Number: 152570-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR