3,5-Dibromo-2-ethoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

75042-500MG 75042-1G 75042-5G
77338-978EA 538.69 USD
77338-978 77358-268 77382-170
3,5-Dibromo-2-ethoxybenzoic acid ≥95%
3,5-Dibromo-2-ethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈Br₂O₃
MW: 323.966 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02729213
CAS Number: 633322-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR