(2-Methoxy-6-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

74907-500MG 74907-1G 74907-5G 74907-25G
77334-086EA 225.57 USD
77334-086 77358-266 77370-094 77390-930
(2-Methoxy-6-(trifluoromethyl)phenyl)(methyl)sulfane ≥95%
(2-Methoxy-6-(trifluoromethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₉F₃OS
MW: 222.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32265516
CAS Number: 2901106-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR