2-(4-Fluorophenyl)-5-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

74898-500MG 74898-1G 74898-5G
77337-232EA 408.91 USD
77337-232 77358-264 77382-168
2-(4-Fluorophenyl)-5-iodopyridine ≥97%
2-(4-Fluorophenyl)-5-iodopyridine

Precaución: for Research Use Only

Formula: C₁₁H₇FIN
Storage Temperature: Ambient
MDL Number: MFCD14699777
CAS Number: 1214348-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR