3,5-Dibromo-2-isopropoxy-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

74847-1G 74847-5G 74847-25G
77341-528EA 91.67 USD
77341-528 77370-090 77382-164
3,5-Dibromo-2-isopropoxy-1,1'-biphenyl ≥97%
3,5-Dibromo-2-isopropoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₄Br₂O
Storage Temperature: Ambient
MDL Number: MFCD34765631
CAS Number: 2807446-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR