3,5-Dibromo-2-methoxy-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

74845-1G 74845-5G 74845-25G
77341-254EA 78.28 USD
77341-254 77370-086 77382-160
3,5-Dibromo-2-methoxy-1,1'-biphenyl ≥95%
3,5-Dibromo-2-methoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀Br₂O
MW: 342.026 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11870634
CAS Number: 1823245-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR