1-Bromo-2-chloro-4-fluoro-5-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

74844-500MG 74844-1G 74844-5G 74844-25G
77334-082EA 163.77 USD
77334-082 77346-132 77370-084 77390-926
1-Bromo-2-chloro-4-fluoro-5-(methoxymethyl)benzene ≥97%
1-Bromo-2-chloro-4-fluoro-5-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₇BrClFO
MW: 253.496 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765629
CAS Number: 2901082-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR