4-(4,4-Dimethylcyclohexyl)phenol ≥97%

Supplier: AOBChem USA

Warning

74827-500MG 74827-1G 74827-25G 74827-5G
77334-080EA 293.55 USD
77334-080 77358-260 77390-924 77382-158
4-(4,4-Dimethylcyclohexyl)phenol ≥97%
4-(4,4-Dimethylcyclohexyl)phenol

Precaución: for Research Use Only

Formula: C₁₄H₂₀O
MW: 204.308 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18452029
CAS Number: 313511-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR