(5-Chloro-3-fluoro-2-isopropoxyphenyl)(methyl)sulfane ≥97%
Supplier: AOBChem USA
|
Warning
|
74813-500MG
74813-1G
74813-5G
74813-25G
77334-076EA
98.88
USD
77334-076
77342-732
77370-078
77382-154
(5-Chloro-3-fluoro-2-isopropoxyphenyl)(methyl)sulfane ≥97%
(5-Chloro-3-fluoro-2-isopropoxyphenyl)(methyl)sulfane
Precaución: for Research Use Only
Formula:
C₁₀H₁₂ClFOS MW: 234.718 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34765617 CAS Number: 2901105-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...