(5-Ethyl-2-fluoro-3-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

74800-500MG 74800-1G 74800-5G
77335-948EA 369.77 USD
77335-948 77358-256 77382-152
(5-Ethyl-2-fluoro-3-iodophenyl)(methyl)sulfane ≥97%
(5-Ethyl-2-fluoro-3-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₀FIS
Storage Temperature: Ambient
MDL Number: MFCD34765613
CAS Number: 2707592-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR