1-(Benzyloxy)-5-bromo-2-chloro-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

74736-500MG 74736-1G 74736-5G
77337-738EA 439.81 USD
77337-738 77358-250 77382-146
1-(Benzyloxy)-5-bromo-2-chloro-4-(trifluoromethyl)benzene ≥97%
1-(Benzyloxy)-5-bromo-2-chloro-4-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₉BrClF₃O
MW: 365.573 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765596
CAS Number: 2901099-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR