1-Bromo-4-chloro-5-isopropoxy-2-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

74733-500MG 74733-1G 74733-5G
77339-814EA 649.93 USD
77339-814 77358-248 77382-144
1-Bromo-4-chloro-5-isopropoxy-2-(trifluoromethyl)benzene ≥95%
1-Bromo-4-chloro-5-isopropoxy-2-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₉BrClF₃O
MW: 317.53 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765593
CAS Number: 2901105-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR