1-Bromo-4-chloro-5-ethoxy-2-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

74730-500MG 74730-1G 74730-5G
77337-898EA 450.11 USD
77337-898 77358-246 77382-142
1-Bromo-4-chloro-5-ethoxy-2-(trifluoromethyl)benzene ≥97%
1-Bromo-4-chloro-5-ethoxy-2-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₇BrClF₃O
Storage Temperature: Ambient
MDL Number: MFCD34765590
CAS Number: 2384501-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR