1-Bromo-2,3-dichloro-4-isopropoxybenzene

Supplier: AOBChem USA

Warning

74726-500MG 74726-1G 74726-5G 74726-25G
77334-070EA 187.46 USD
77334-070 77358-244 77370-072 77390-918
1-Bromo-2,3-dichloro-4-isopropoxybenzene
1-Bromo-2,3-dichloro-4-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉BrCl₂O
MW: 283.977 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29043913
CAS Number: 1881331-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR