1-Bromo-2,3-dichloro-4-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

74725-500MG 74725-1G 74725-5G 74725-25G
77334-068EA 99.91 USD
77334-068 77342-886 77370-070 77382-138
1-Bromo-2,3-dichloro-4-ethoxybenzene ≥97%
1-Bromo-2,3-dichloro-4-ethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₇BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD21609661
CAS Number: 1253972-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR