2-Bromo-6-chloro-4-isobutylphenol ≥97%

Supplier: AOBChem USA

Warning

74718-500MG 74718-1G 74718-5G 74718-25G
77334-066EA 207.03 USD
77334-066 77358-242 77370-068 77390-916
2-Bromo-6-chloro-4-isobutylphenol ≥97%
2-Bromo-6-chloro-4-isobutylphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrClO
MW: 263.559 g/mol
Storage Temperature: Ambient
CAS Number: 2918947-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR