(3-Bromo-5-propoxyphenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

74705-500MG 74705-1G 74705-5G
77338-930EA 531.48 USD
77338-930 77358-240 77382-136
(3-Bromo-5-propoxyphenyl)trimethylsilane ≥97%
(3-Bromo-5-propoxyphenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₁₂H₁₉BrOSi
MW: 287.268 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765582
CAS Number: 2901082-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR