(3-Bromo-5-propoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

74704-500MG 74704-1G 74704-5G 74704-25G
77334-064EA 279.13 USD
77334-064 77358-238 77370-066 77390-914
(3-Bromo-5-propoxyphenyl)(methyl)sulfane ≥95%
(3-Bromo-5-propoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrOS
Storage Temperature: Ambient
MDL Number: MFCD34765581
CAS Number: 2901085-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR