1-(2,6-Difluoro-4-isopropoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

74702-500MG 74702-1G 74702-25G 74702-5G
77334-062EA 301.79 USD
77334-062 77358-234 77390-912 77382-132
1-(2,6-Difluoro-4-isopropoxyphenyl)ethanone ≥97%
1-(2,6-Difluoro-4-isopropoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₂
MW: 214.209 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765579
CAS Number: 2901105-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR