1,3-Difluoro-2-iodo-5-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

74700-500MG 74700-1G 74700-5G 74700-25G
77334-060EA 178.19 USD
77334-060 77346-626 77370-064 77390-910
1,3-Difluoro-2-iodo-5-isopropoxybenzene ≥97%
1,3-Difluoro-2-iodo-5-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉F₂IO
Storage Temperature: Ambient
MDL Number: MFCD34765577
CAS Number: 2901086-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR