2-Bromo-1,3-difluoro-5-isopropoxybenzene ≥95%

Supplier: AOBChem USA

Warning

74699-500MG 74699-1G 74699-5G 74699-25G
77334-058EA 157.59 USD
77334-058 77345-676 77370-062 77390-908
2-Bromo-1,3-difluoro-5-isopropoxybenzene ≥95%
2-Bromo-1,3-difluoro-5-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉BrF₂O
MW: 251.068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765576
CAS Number: 2901066-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR